missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzoic acid, 95%, ACROS Organics™

Brand:  Acros Organics 278320010

Code : 29

Additional Details : CAS Number : 552-16-9 Weight : 1.05000kg

Packaging Glass bottle
Quantity 1kg

 View more versions of this product

Product Code. 10006003

  • 192.00 € / 1kg

Estimated Shipment: 06-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

552-16-9
167.12
SLAMLWHELXOEJZ-UHFFFAOYSA-N
11087
C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-]
C7H5NO4
MFCD00007137
2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid
CHEBI:25620

Specifications

2-Nitrobenzoic acid
Glass bottle
141.0°C to 148.0°C
85-90%
85.0
94% min. (GC)
O2NC6H4COOH
09, 370
14, 6589
Solubility in water: 6.8g/l. Other solubilities: 1g/3mL alcohol- 1g/2.5mL acetone,1g/220mL chloroform- 1g/2.5mL methanol,1g/4.5mL ether,slightly soluble in benzene,cs2,petroleum ether
C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-]
11087
167.12
95%
Authentic
White
1kg
552-16-9
100.0
C7H5NO4
MFCD00007137
13,231
2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid
SLAMLWHELXOEJZ-UHFFFAOYSA-N
167.12
CHEBI:25620
Fine Powder
Videos
SDS Documents
Certificates