missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrobenzonitrile, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 110.8°C to 111.8°C |
Color | Yellow |
Boiling Point | 165.0°C (16.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | O2NC6H4CN |
Quantity | 100 g |
Show More |
Safety and Handling
Product Identifier
- 2-Nitrobenzonitrile
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H300-Fatal if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'