missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-4229313
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
612-24-8
C7H4N2O2
148.12
MFCD00007044
SWBDKCMOLSUXRH-UHFFFAOYSA-N
o-nitrobenzonitrile, o-cyanonitrobenzene, benzonitrile, o-nitro, benzonitrile, 2-nitro, 2-cyanonitrobenzene, nitrobenzonitrile, unii-drc6u29fci, ccris 2326, 1-nitro-2-cyanobenzene, 2-nitrobenzenecarbonitrile
11922
2-nitrobenzonitrile
[O-][N+](=O)C1=CC=CC=C1C#N
This item is not returnable. View return policy
612-24-8
C7H4N2O2
148.12
MFCD00007044
SWBDKCMOLSUXRH-UHFFFAOYSA-N
o-nitrobenzonitrile, o-cyanonitrobenzene, benzonitrile, o-nitro, benzonitrile, 2-nitro, 2-cyanonitrobenzene, nitrobenzonitrile, unii-drc6u29fci, ccris 2326, 1-nitro-2-cyanobenzene, 2-nitrobenzenecarbonitrile
11922
2-nitrobenzonitrile
[O-][N+](=O)C1=CC=CC=C1C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 110.8°C to 111.8°C
Color Yellow
Boiling Point 165.0°C (16.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CN
Quantity 10 g
Beilstein 09,374
Solubility Information Solubility in water: insoluble
Formula Weight 148.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrobenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'