missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanone, 99+%, purified by redistillation, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4812841
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass bottle
107-87-9
C5H10O
86.13
XNLICIUVMPYHGG-UHFFFAOYSA-N
2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone
7895
CHEBI:16472
pentan-2-one
CCCC(=O)C
107-87-9
C5H10O
86.13
XNLICIUVMPYHGG-UHFFFAOYSA-N
2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone
7895
CHEBI:16472
pentan-2-one
CCCC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -78.0°C
Density 0.8090g/mL
Boiling Point 100.0°C to 103.0°C
Flash Point 7°C
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula CH3CO(CH2)CH3
Refractive Index 1.3890 to 1.3910
Quantity 100 mL
Beilstein 01,676
Merck Index 15,6187
Specific Gravity 0.809
Solubility Information Solubility in water: 43g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.5 mPa.s (20°C)
Formula Weight 86.13
Percent Purity 99+%
Chemical Name or Material 2-Pentanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'