missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthiazole, 98%, Thermo Scientific Chemicals

Catalog Number 15264228
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1826-11-5
C9H7NS
161.23
WYKHSBAVLOPISI-UHFFFAOYSA-N
2-phenylthiazole, thiazole, 2-phenyl, phenylthiazole, 2-phenyl thiazole, #, ksc174s8f, tpc-i164
547494
2-phenyl-1,3-thiazole
C1=CC=C(C=C1)C2=NC=CS2
This item is not returnable. View return policy
1826-11-5
C9H7NS
161.23
WYKHSBAVLOPISI-UHFFFAOYSA-N
2-phenylthiazole, thiazole, 2-phenyl, phenylthiazole, 2-phenyl thiazole, #, ksc174s8f, tpc-i164
547494
2-phenyl-1,3-thiazole
C1=CC=C(C=C1)C2=NC=CS2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 161.23
Percent Purity 98%
Chemical Name or Material 2-Phenylthiazole
compliance-icons
Product Identifier
  • 2-Phenylthiazole, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'