missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, 99+%, Thermo Scientific Chemicals

Catalog Number p-7045299
Click to view available options
Quantity:
1000 mL
2500 mL
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

2-Propanol is used as a solvent for gums, resins, alkaloids and essential oils, as a denaturant for ethyl alcohol and is used in lotions and cosmetics. It is used in keyboard, LCD and laptop cleaning, as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It acts as hydride source in the Meerwein-Ponndorf-Verley reduction. It plays an important role in DNA extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Propanol is used as a solvent for gums, resins, alkaloids and essential oils, as a denaturant for ethyl alcohol and is used in lotions and cosmetics. It is used in keyboard, LCD and laptop cleaning, as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It acts as hydride source in the Meerwein-Ponndorf-Verley reduction. It plays an important role in DNA extraction.

Solubility
Miscible with water, ethanol, acetone, chloroform, methanol, diethyl ether, n-octanol and benzene.

Notes
It is a flammable chemical compound and is hygroscopic in nature. Heat and sunlight can contribute to its instability.
TRUSTED_SUSTAINABILITY
Melting Point -90°C
UN Number UN1219
Beilstein 635639
Merck Index 14,5208
Solubility Information Miscible with water,ethanol,acetone,chloroform,methanol,diethyl ether,n-octanol and benzene.
Formula Weight 60.1
Percent Purity ≥99%
Density 0.785
Boiling Point 82°C to 83°C
Quantity 1000 mL
Flash Point 12°C (53°F)
Odor Alcohol-like
Refractive Index 1.377
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'