Learn More
2-Propanol, ACS, 99.5% min, Thermo Scientific Chemicals
Description
It acts as a solvent for gums, resins, alkaloids and essential oils. It is used in cosmetic lotions. Isopropyl alcohol is used as a cleansing agent for keyboards, LCD displays and laptops. It is used as a wetting agent in the fountain solution used in lithographic printing, and is often used as a solvent for french polishing shellac, which is used in cabinet making. It act as a hydride source in the Meerwein-Ponndorf-Verley reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -90°C |
UN Number | UN1219 |
Beilstein | 635639 |
Merck Index | 14,5208 |
Solubility Information | Miscible with water,alcohol,ether,chloroform,acetone and benzene. |
Formula Weight | 60.1 |
Percent Purity | ≥99.5% |
Grade | ACS |
Density | 0.785 |
Boiling Point | 82°C to 83°C |
Show More |
RUO – Research Use Only