Learn More
Thermo Scientific Chemicals 2-Pyrrolidone-5-carboxylic acid, sodium salt, 50 wt.% solution in water
Brand: Thermo Scientific Chemicals 232921000
Code : 29
Additional Details : CAS Number : 54571-67-4 Weight : 0.12700kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Pyrrolidone-5-carboxylic acid, sodium salt | |
50 wt. % Solution in Water | |
47.5 | |
C5H6NNaO3 | |
1.27g/mL | |
CRPCXAMJWCDHFM-DFWYDOINSA-M | |
sodium;5-oxopyrrolidine-2-carboxylate | |
23666346 | |
47.5 to 52.5% | |
6.8 to 7.4 | |
Solution |
Colorless | |
54571-67-4,7732-18-5 | |
52.5 | |
MFCD00066984 | |
sodium 5-oxopyrrolidine-2-carboxylate, sodium 5-oxo-dl-prolinate, monosodium 5-oxoproline, sodium 2-pyrrolidone-5-carboxylate, pca soda, proline, 5-oxo-, sodium salt 1:1, 2-pyrrolidone-5-carboxylic acid sodium salt, nalidone, 5-oxo-dl-proline, monosodium salt, dl-proline, 5-oxo-, monosodium salt | |
[Na+].[O-]C(=O)[C@@H]1CCC(=O)N1 | |
151.10 | |
151.1 | |
Glass bottle | |
100 mL |
RUO – Research Use Only