missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, 98%, ACROS Organics™

Brand:  Acros Organics 439160010

Code : 29

Additional Details : CAS Number : 98015-45-3 Weight : 0.05095kg Transport : UN number : 2920 Chem class : 8 Pack group : II


Packaging Glass bottle
Quantity 1mL

 View more versions of this product

Product Code. 11945711

  • 92.83 € / 1mL

Estimated Shipment
25-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

98015-45-3
274.39
bemp, bemp phosphazene, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, unii-df15j146qi, 2-tert-butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2??-diazaphosphinan-2-amine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2, 2-t-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-1,3,2-diazaphosphorinane
2-tert-butylimino-N,N-diethyl-1,3-dimethyl-1,3,2$l^{5}-diazaphosphinan-2-amine
C13H31N4P
VSCBATMPTLKTOV-UHFFFAOYSA-N
3513851
CCN(CC)P1(=NC(C)(C)C)N(CCCN1C)C

Specifications

2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine
53°C
Glass bottle
74°C (0.1mmHg)
1mL
97.5
97.5% min. (GC)
bemp, bemp phosphazene, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, unii-df15j146qi, 2-tert-butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2??-diazaphosphinan-2-amine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2, 2-t-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-1,3,2-diazaphosphorinane
CCN(CC)P1(=NC(C)(C)C)N(CCCN1C)C
274.39
274.39
98%
0.9480g/mL
Authentic
0.948
Colorless
98015-45-3
100.0
C13H31N4P
VSCBATMPTLKTOV-UHFFFAOYSA-N
2-tert-butylimino-N,N-diethyl-1,3-dimethyl-1,3,2$l^{5}-diazaphosphinan-2-amine
3513851
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents