missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ASTM Certificate, Pore Sizes: 10mm and Larger

Brand:  RETSCH 60.141.075000

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 11964682

  • 148.00 € / Each


Estimated Shipment: 10-08-2021
Add to basket

Description

Description

Specifications

Specifications

For separation, fractioning, particle size determination
Stainless Steel
1Each
Test Sieve
Stainless Steel
25.4mm
203.2mm
Stainless Steel
75mm
ASTM E 11
Shaker AS 300
Videos
SDS
Documents
Certificates