missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ISO Certificate, Micrometer Pore Sizes

Brand:  RETSCH 60.140.000063

Code : X3

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 11954452

  • 111.00 € / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
25.4mm
For separation, fractioning, particle size determination
203.2 mm
63μm
ISO 3310/1
Shaker AS 300
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers