Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ISO Certificate, Micrometer Pore Sizes
Brand:  RETSCH 60.140.000450
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 11974472
  • 111.00 € / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Specifications
Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
25.4mm
For separation, fractioning, particle size determination
203.2 mm
450μm
ISO 3310/1
Shaker AS 300