missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ISO Certificate, Pore Sizes: 10mm and Larger
Brand: RETSCH 60.140.080000
Additional Details : Weight : 0.20000kg
Specifications
For separation, fractioning, particle size determination | |
Stainless Steel | |
1Each | |
Test Sieve | |
Stainless Steel | |
25.4mm |
203.2mm | |
Stainless Steel | |
80mm | |
ISO 3310/1 | |
Shaker AS 300 |