missing translation for 'onlineSavingsMsg'
Learn More

(2R,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, 98% ee, Thermo Scientific Chemicals

Catalog Number 10028781
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
321744-26-7
C12H21NO5
259.30
MFCD02259744
RNMVWSAJMIKMDY-DTWKUNHWSA-N
2734504
1-O-tert-butyl 2-O-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate
COC(=O)[C@H]1C[C@@H](O)CCN1C(=O)OC(C)(C)C
This item is not returnable. View return policy
321744-26-7
C12H21NO5
259.30
MFCD02259744
RNMVWSAJMIKMDY-DTWKUNHWSA-N
2734504
1-O-tert-butyl 2-O-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate
COC(=O)[C@H]1C[C@@H](O)CCN1C(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Amber to Colorless
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid methyl ester, 2r,4s-1-tert-butyl 2-methyl 4-hydroxypiperidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid, 2r,4s-1-tert-butyl2-methyl4-hydroxypiperidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-cis-4-hydroxy-l-pipecolinic acid methyl ester, 1-o-tert-butyl 2-o-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, o1-tert-butyl o2-methyl 2r,4s-4-oxidanylpiperidine-1,2-dicarboxylate, tert-butyl 4s,2r-4-hydroxy-2-methoxycarbonyl piperidinecarboxylate, 2r-4beta-hydroxypiperidine-1,2beta-dicarboxylic acid 1-tert-butyl 2-methyl ester
Formula Weight 259.3
Percent Purity 95%,98% ee
Physical Form Viscous Liquid or Waxy Mass
Chemical Name or Material (2R, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'