missing translation for 'onlineSavingsMsg'
Learn More

2xYT Agar (Powder/Molecular Genetics), Fisher BioReagents

Brand:  Fisher Bioreagents BP2466-500

Code : 32

Additional Details : CAS Number : 9002-18-0 Weight : 0.55000kg

 View more versions of this product

Product Code. 10041793

  • 86.98 € / 500g

Estimated Shipment: 22-04-2022
Add to basket

Description

Description

Specifications

Specifications

Poly Bottle
Powder
500 g
2xyt Agar
Molecular Genetics
Videos
SDS Documents
Certificates
Special Offers

Special Offers