missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde, 90%, Thermo Scientific™

Catalog Number p-3522468
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
179056-79-2
C11H10N2O
186.21
MFCD08669912
SSAPUQDWKCUIAR-UHFFFAOYSA-N
10511755
3-(1-methylpyrazol-3-yl)benzaldehyde
CN1C=CC(=N1)C1=CC(C=O)=CC=C1
This item is not returnable. View return policy
179056-79-2
C11H10N2O
186.21
MFCD08669912
SSAPUQDWKCUIAR-UHFFFAOYSA-N
10511755
3-(1-methylpyrazol-3-yl)benzaldehyde
CN1C=CC(=N1)C1=CC(C=O)=CC=C1
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-1-methyl-1h-pyrazol-3-yl benzaldehyde, 3-1-methylpyrazol-3-yl benzaldehyde, benzaldehyde,3-1-methyl-1h-pyrazol-3-yl, benzaldehyde, 3-1-methyl-1h-pyrazol-3-yl, 3-3-formylphenyl-1-methyl-1h-pyrazole
Formula Weight 186.21
Percent Purity 90%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'