missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes
Use round-bottom 3.3 Borosilicate Glass Test Tubes for laboratory needs when heat resistance is critical. These disposable test tubes are thick-walled and autoclavable.
Brand: DWK Life Sciences A618020001221
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Disposable
- Borosilicate glass
- Excellent resistance to temperature changes
- Autoclavable
Specifications
Test Tube | |
Round bottom | |
100Pack |
20 mm | |
180 mm |