missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes

Use round-bottom 3.3 Borosilicate Glass Test Tubes for laboratory needs when heat resistance is critical. These disposable test tubes are thick-walled and autoclavable.

Brand:  DWK Life Sciences A620025001221

Code : A0

Additional Details : Weight : 0.80000kg

Product Code. 11832363

  • 39.00 € / Pack of 50
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Description

Description

  • Disposable
  • Borosilicate glass
  • Excellent resistance to temperature changes
  • Autoclavable
Specifications

Specifications

Test Tube
Round bottom
50Pack
25 mm
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers