missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylbutan-2-one oxime, 98%, Thermo Scientific™

Catalog Number 11945661
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2475-93-6
C6H13NO
115.18
UNSDDJQNHCSVSW-FNORWQNLSA-N
pinacolone oxime, 3,3-dimethyl-2-butanone oxime, 2-butanone,3-dimethyl-, oxime, 2-hydroxyimino-3,3-dimethylbutane, pinakolinoxim, 2e-n-hydroxy-3,3-dimethylbutan-2-imine
9561079
(NE)-N-(3,3-dimethylbutan-2-ylidene)hydroxylamine
CC(=NO)C(C)(C)C
This item is not returnable. View return policy
2475-93-6
C6H13NO
115.18
UNSDDJQNHCSVSW-FNORWQNLSA-N
pinacolone oxime, 3,3-dimethyl-2-butanone oxime, 2-butanone,3-dimethyl-, oxime, 2-hydroxyimino-3,3-dimethylbutane, pinakolinoxim, 2e-n-hydroxy-3,3-dimethylbutan-2-imine
9561079
(NE)-N-(3,3-dimethylbutan-2-ylidene)hydroxylamine
CC(=NO)C(C)(C)C
CAS Min % 97.5
CAS Max % 100.0
Melting Point 75°C to 77°C
Color White
Boiling Point 171°C to 172°C (748.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 115.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3-Dimethylbutan-2-one oxime
Show More Show Less
compliance-icons
Product Identifier
  • 3,3-Dimethylbutan-2-one oxime
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333-If skin irritation or rash occurs:.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'