missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)propionic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-3770778
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1643-30-7
MFCD01310793
NCSTWHYWOVZDOC-UHFFFAOYSA-N
3-4-bromophenyl propionic acid, 3-4-bromophenyl propanoic acid, benzenepropanoic acid, 4-bromo, 3-4-bromo-phenyl-propionic acid, p-bromohydrocinnamic acid, 3-4-bromophenyl-propionic acid, 3-4-bromophenyl propanoicacid, pubchem19769, 4-bromohydrocinnamic acid, acmc-1c0vm
2735609
3-(4-bromophenyl)propanoic acid
C1=CC(=CC=C1CCC(=O)O)Br
This item is not returnable. View return policy
1643-30-7
MFCD01310793
NCSTWHYWOVZDOC-UHFFFAOYSA-N
3-4-bromophenyl propionic acid, 3-4-bromophenyl propanoic acid, benzenepropanoic acid, 4-bromo, 3-4-bromo-phenyl-propionic acid, p-bromohydrocinnamic acid, 3-4-bromophenyl-propionic acid, 3-4-bromophenyl propanoicacid, pubchem19769, 4-bromohydrocinnamic acid, acmc-1c0vm
2735609
3-(4-bromophenyl)propanoic acid
C1=CC(=CC=C1CCC(=O)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 133°C to 136°C
Color Beige or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H9BrO2
Linear Formula BrC6H4(CH2)2COOH
Quantity 1 g
Molecular Weight (g/mol) 229.1
Formula Weight 229.1
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Bromophenyl)propionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-(4-Bromophenyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'