missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenoxy)pentane-2,4-dione, Tech., Thermo Scientific™

Catalog Number p-2699081
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
31168-10-2
C11H11ClO3
226.656
MFCD00204239
QOOLFYCEZZCVSE-UHFFFAOYSA-N
3-4-chlorophenoxy pentane-2,4-dione, 2,4-pentanedione,3-4-chlorophenoxy, 3-4-chlorophenoxy-2,4-pentanedione
2799447
3-(4-chlorophenoxy)pentane-2,4-dione
CC(=O)C(C(=O)C)OC1=CC=C(C=C1)Cl
This item is not returnable. View return policy
31168-10-2
C11H11ClO3
226.656
MFCD00204239
QOOLFYCEZZCVSE-UHFFFAOYSA-N
3-4-chlorophenoxy pentane-2,4-dione, 2,4-pentanedione,3-4-chlorophenoxy, 3-4-chlorophenoxy-2,4-pentanedione
2799447
3-(4-chlorophenoxy)pentane-2,4-dione
CC(=O)C(C(=O)C)OC1=CC=C(C=C1)Cl
Linear Formula ClC6H4OCHC(O)CH32
Formula Weight 226.66
Percent Purity Tech.
Grade Technical
Quantity 1 g
Chemical Name or Material 3-(4-Chlorophenoxy)pentane-2, 4-dione
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'