missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenoxyacetic acid, 96%, ACROS Organics™

Brand:  Acros Organics 152980100

Code : 29

Additional Details : CAS Number : 588-22-7 Weight : 0.06000kg


Quantity 10g

 View more versions of this product

Product Code. 11935121

  • 163.00 € / 10g

Estimated Shipment: 05-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

588-22-7
221.03
SNYRXHULAWEECU-UHFFFAOYSA-N
11495
2-(3,4-dichlorophenoxy)acetic acid
C8H6Cl2O3
MFCD00004303
3,4-dichlorophenoxyacetic acid, 2-3,4-dichlorophenoxy acetic acid, 3,4-dichlorophenoxy acetic acid, acetic acid, 3,4-dichlorophenoxy, acetic acid,4-dichlorophenoxy, acmc-20anfd, 4-06-00-00954 beilstein handbook reference, 3,4-dichloro-phenoxy-acetic acid, acetic acid,2-3,4-dichlorophenoxy
CHEBI:393747
OC(=O)COC1=CC=C(Cl)C(Cl)=C1

Specifications

3, 4-Dichlorophenoxyacetic acid
White
10g
95.0
96% min. (HPLC)
Cl2C6H3OCH2CO2H
3,4-dichlorophenoxyacetic acid, 2-3,4-dichlorophenoxy acetic acid, 3,4-dichlorophenoxy acetic acid, acetic acid, 3,4-dichlorophenoxy, acetic acid,4-dichlorophenoxy, acmc-20anfd, 4-06-00-00954 beilstein handbook reference, 3,4-dichloro-phenoxy-acetic acid, acetic acid,2-3,4-dichlorophenoxy
OC(=O)COC1=CC=C(Cl)C(Cl)=C1
221.03
CHEBI:393747
Crystalline Powder
Authentic
138.0°C to 142.0°C
588-22-7
100.0
C8H6Cl2O3
MFCD00004303
SNYRXHULAWEECU-UHFFFAOYSA-N
2-(3,4-dichlorophenoxy)acetic acid
11495
221.04
96%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,4-Dichlorophenoxyacetic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates