missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Difluoro-N-(4-methoxybenzyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61737.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416756

  • 99.64 € / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

3,4-Difluoro-N-(4-methoxybenzyl)benzenesulfonamide
313.32
MFCD05107704

Specifications

3,4-Difluoro-N-(4-methoxybenzyl)benzenesulfonamide
Solid
313.32
1g
97%
MFCD05107704
Videos
SDS