missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzylamine, 95%, Thermo Scientific™

Catalog Number p-2690220
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
72235-53-1
C7H7F2N
143.14
MFCD00010145
PHLZUDXEBCQHKM-UHFFFAOYSA-N
123572
(3,4-difluorophenyl)methanamine
C1=CC(=C(C=C1CN)F)F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
72235-53-1
C7H7F2N
143.14
MFCD00010145
PHLZUDXEBCQHKM-UHFFFAOYSA-N
123572
(3,4-difluorophenyl)methanamine
C1=CC(=C(C=C1CN)F)F
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2100g/mL
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CH2NH2
Refractive Index 1.4920 to 1.4940
Quantity 1 g
Specific Gravity 1.21
Synonym 3,4-difluorobenzylamine, 3,4-difluorophenyl methanamine, 3,4-difluorobenzyl amine, benzenemethanamine, 3,4-difluoro, 1-3,4-difluorophenyl methanamine, 3,4-difluorophenyl methylamine, 3, 4-difluorobenzylamine, pubchem1918, acmc-209omb, 3,4-difluoro-benzylamine
Formula Weight 143.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3, 4-Difluorobenzylamine, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'