Learn More
(3,4-Dimethoxyphenyl)acetonitrile, 99+%, ACROS Organics™
Brand: Acros Organics 155260250
Code : 29
Additional Details : CAS Number : 93-17-4 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 25g |
Chemical Identifiers
93-17-4 | |
177.2 | |
ASLSUMISAQDOOB-UHFFFAOYSA-N | |
66727 | |
COC1=C(C=C(C=C1)CC#N)OC |
C10H11NO2 | |
MFCD00001911 | |
3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile | |
2-(3,4-dimethoxyphenyl)acetonitrile |
Specifications
(3, 4-Dimethoxyphenyl)acetonitrile | |
Authentic | |
Glass bottle | |
171°C to 178°C (10.0mmHg) | |
62°C to 65°C | |
93-17-4 | |
100.0 | |
C10H11NO2 | |
MFCD00001911 | |
3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile | |
ASLSUMISAQDOOB-UHFFFAOYSA-N | |
2-(3,4-dimethoxyphenyl)acetonitrile | |
66727 | |
Crystals or Crystalline Needles |
250°C | |
0.5% max. (vacuum) | |
0.1% max. | |
White | |
25g | |
99.0 | |
99+% | |
(CH3O)2C6H3CH2CN | |
10, I, 198 | |
Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,chloroform and,ethanol | |
COC1=C(C=C(C=C1)CC#N)OC | |
177.2 | |
177.2 | |
99+% |
Safety and Handling
- (3,4-Dimethoxyphenyl)acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.