missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific Chemicals

Catalog Number 10023590
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
32247-96-4
C9H5BrF6
307.03
MFCD00009905
ATLQGZVLWOURFU-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
122573
1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F
This item is not returnable. View return policy
32247-96-4
C9H5BrF6
307.03
MFCD00009905
ATLQGZVLWOURFU-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
122573
1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Density 1.6700g/mL
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CH2Br
Refractive Index 1.4440 to 1.446
Quantity 1 g
Specific Gravity 1.67
Formula Weight 307.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'