missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(5-Chloro-2-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56606.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15472796

  • 671.00 € / 1g


Estimated Shipment date
from Supplier 22-05-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665222

Specifications

3-(5-Chloro-2-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid
MFCD27665222
Solid
1g
307.51
97%
Videos
SDS