missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-102962
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
1620-98-0
MFCD00008826
DOZRDZLFLOODMB-UHFFFAOYSA-N
3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehyde
73219
3,5-ditert-butyl-4-hydroxybenzaldehyde
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O
This item is not returnable. View return policy
1620-98-0
MFCD00008826
DOZRDZLFLOODMB-UHFFFAOYSA-N
3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehyde
73219
3,5-ditert-butyl-4-hydroxybenzaldehyde
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 186.0°C to 190.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C15H22O2
Quantity 25 g
Molecular Weight (g/mol) 234.34
Formula Weight 234.34
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Di-tert-butyl-4-hydroxybenzaldehyde, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'