missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromopyridine, 98+%

Product Code. 10523051 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10523051

Brand: Thermo Scientific Acros 330130100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-92-3 | C5H3Br2N | 236.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-92-3
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.89
MDL Number MFCD00014634
InChI Key SOSPMXMEOFGPIM-UHFFFAOYSA-N
Synonym pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln
PubChem CID 69369
ChEBI CHEBI:51593
IUPAC Name 3,5-dibromopyridine
SMILES BrC1=CC(Br)=CN=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 110°C to 114°C
Color White to Beige
Flash Point >115°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol
Formula Weight 236.91
Percent Purity 98+%
Physical Form Needle-like Crystals
Chemical Name or Material 3, 5-Dibromopyridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.