missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-1,2,4-thiadiazole, 97%, Thermo Scientific Chemicals

Catalog Number p-4510701
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2254-88-8
C2Cl2N2S
155.01
CEUHPOVLEQUFCC-UHFFFAOYSA-N
2725055
3,5-dichloro-1,2,4-thiadiazole
C1(=NSC(=N1)Cl)Cl
This item is not returnable. View return policy
2254-88-8
C2Cl2N2S
155.01
CEUHPOVLEQUFCC-UHFFFAOYSA-N
2725055
3,5-dichloro-1,2,4-thiadiazole
C1(=NSC(=N1)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point 165.0°C to 167.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.5600
Quantity 1 g
Synonym dichloro-1,2,4-thiadiazole, 3,5-dichloro-1,2,4 thiadiazole, 1,2,4-thiadiazole, 3,5-dichloro, 1,2,4-thiadiazole,3,5-dichloro, 3,5-dichloro-1,2,4 thiadiazol
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 155.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3,5-Dichloro-1,2,4-thiadiazole
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dichloro-1,2,4-thiadiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'