missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-2,4,6-trifluoropyridine, 98%, ACROS Organics™

Brand:  Acros Organics 311400050

Code : 29

Additional Details : CAS Number : 1737-93-5 Weight : 0.05500kg Transport : UN number : 3382 Chem class : 6.1 Pack group : I

Quantity 5g

 View more versions of this product

Product Code. 10003630

  • 20.10 € / 5g

Estimated Shipment
23-10-2020

Please to purchase this item. Need a web account? Register with us today!


Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

1737-93-5
201.96
2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
3,5-dichloro-2,4,6-trifluoropyridine
C5Cl2F3N
PKSORSNCSXBXOT-UHFFFAOYSA-N
61273
C1(=C(C(=NC(=C1Cl)F)F)Cl)F

Specifications

3,5-Dichloro-2,4,6-trifluoropyridine
113°C
1.622
24.0°C to 26.0°C
1737-93-5
C5Cl2F3N
PKSORSNCSXBXOT-UHFFFAOYSA-N
3,5-dichloro-2,4,6-trifluoropyridine
61273
98%
1.6220g/mL
Glass bottle
159.0°C to 160.0°C
5g
97.5% min. (GC)
2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
C1(=C(C(=NC(=C1Cl)F)F)Cl)F
201.96
201.96
Videos
SDS Documents