Learn More
3-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-5-hydroxybenzoic acid, ≥97%, Maybridge™
Brand: Maybridge MO07963DA
Code : 31
Additional Details : CAS Number : 176442-21-0 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
176442-21-0 | |
375.38 | |
WGBZDECNYLZYRE-UHFFFAOYSA-N | |
11057866 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC(=CC(=C4)C(=O)O)O |
C22H17NO5 | |
MFCD02682203 | |
3-9h-fluoren-9-ylmethoxy carbonyl amino-5-hydroxybenzoic acid, 3-9h-fluoren-9-yl methoxy carbonyl amino-5-hydroxybenzoic acid, 3-amino-5-hydroxybenzoic acid,n-fmoc protected, 3-hydroxy-5-9h-fluorene-9-yl methoxy carbonyl amino benzoic acid, benzoic acid,3-9h-fluoren-9-ylmethoxy carbonyl amino-5-hydroxy | |
3-(9H-fluoren-9-ylmethoxycarbonylamino)-5-hydroxybenzoic acid |
Specifications
3-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-5-hydroxybenzoic acid | |
1g | |
97.0 | |
C22H17NO5 | |
3-9h-fluoren-9-ylmethoxy carbonyl amino-5-hydroxybenzoic acid, 3-9h-fluoren-9-yl methoxy carbonyl amino-5-hydroxybenzoic acid, 3-amino-5-hydroxybenzoic acid,n-fmoc protected, 3-hydroxy-5-9h-fluorene-9-yl methoxy carbonyl amino benzoic acid, benzoic acid,3-9h-fluoren-9-ylmethoxy carbonyl amino-5-hydroxy | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC(=CC(=C4)C(=O)O)O | |
375.38 | |
375.38 |
Amber Glass Bottle | |
176442-21-0 | |
97+% | |
MFCD02682203 | |
WGBZDECNYLZYRE-UHFFFAOYSA-N | |
3-(9H-fluoren-9-ylmethoxycarbonylamino)-5-hydroxybenzoic acid | |
11057866 | |
97% |