missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2,4-dimethylpyrrole, 98%, ACROS Organics™

Brand:  Acros Organics 102510500

Code : 29

Additional Details : CAS Number : 2386-25-6 Weight : 0.10000kg


Packaging Glass bottle
Quantity 50g

 View more versions of this product

Product Code. 11966591

  • 496.00 € / 50g

Estimated Shipment
26-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2386-25-6
137.18
3-acetyl-2,4-dimethylpyrrole, 2,4-dimethyl-3-acetylpyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl ethanone, 2,4-dimethylpyrrol-3-yl methyl ketone, ethanone, 1-2,4-dimethyl-1h-pyrrol-3-yl, ketone, 2,4-dimethylpyrrol-3-yl methyl, 1-2,4-dimethyl-1h-pyrrol-3-yl ethan-1-one, 2,4-dimethyl-3-acetyl-pyrrole, 3-acetyl-2,5-dimethyl-pyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl-ethanone
1-(2,4-dimethyl-1H-pyrrol-3-yl)ethanone
C8H11NO
VGZCKCJMYREIKA-UHFFFAOYSA-N
15163
CC1=CNC(=C1C(=O)C)C

Specifications

3-Acetyl-2, 4-dimethylpyrrole
Glass bottle
136.0°C to 139.0°C
2386-25-6
C8H11NO
VGZCKCJMYREIKA-UHFFFAOYSA-N
1-(2,4-dimethyl-1H-pyrrol-3-yl)ethanone
15163
Powder or Crystals
Authentic
Brown to Yellow
50g
97.5% min. (HPLC)
3-acetyl-2,4-dimethylpyrrole, 2,4-dimethyl-3-acetylpyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl ethanone, 2,4-dimethylpyrrol-3-yl methyl ketone, ethanone, 1-2,4-dimethyl-1h-pyrrol-3-yl, ketone, 2,4-dimethylpyrrol-3-yl methyl, 1-2,4-dimethyl-1h-pyrrol-3-yl ethan-1-one, 2,4-dimethyl-3-acetyl-pyrrole, 3-acetyl-2,5-dimethyl-pyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl-ethanone
CC1=CNC(=C1C(=O)C)C
137.18
137.18
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Acetyl-2,4-dimethylpyrrole

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents