Learn More
3-Amino-5-bromo-1H-indazole, 95%, ACROS Organics™
Brand: Acros Organics 435760010
Code : 29
Additional Details : CAS Number : 61272-71-7 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
61272-71-7 | |
212.05 | |
OMPYFDJVSAMSMA-UHFFFAOYSA-N | |
817910 | |
C1=CC2=C(C=C1Br)C(=NN2)N |
C7H6BrN3 | |
MFCD03426696 | |
3-amino-5-bromo-1h-indazole, 3-amino-5-bromoindazole, 5-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 5-bromo, 5-bromo-1h-indazole-3-ylamine, pubchem18573, 3-amino-5-bromo-indazole, acmc-1b3h9, 5-bromo-1h-indazole-3-amine | |
5-bromo-1H-indazol-3-amine |
Specifications
3-Amino-5-bromo-1H-indazole | |
Glass bottle | |
237°C to 238°C | |
61272-71-7 | |
100.0 | |
C7H6BrN3 | |
3-amino-5-bromo-1h-indazole, 3-amino-5-bromoindazole, 5-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 5-bromo, 5-bromo-1h-indazole-3-ylamine, pubchem18573, 3-amino-5-bromo-indazole, acmc-1b3h9, 5-bromo-1h-indazole-3-amine | |
C1=CC2=C(C=C1Br)C(=NN2)N | |
212.05 | |
212.05 | |
95% |
Authentic | |
Colorless | |
1g | |
94.0 | |
94% min. (HPLC) | |
MFCD03426696 | |
OMPYFDJVSAMSMA-UHFFFAOYSA-N | |
5-bromo-1H-indazol-3-amine | |
817910 | |
Liquid |
Safety and Handling
- 3-Amino-5-bromo-1H-indazole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.