missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-2-chloro-6-fluorotoluene, 97%

36.50 € - 550.00 €

Chemical Identifiers

CAS 203302-92-5
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.469
MDL Number MFCD00236591
InChI Key VHQALVHKNXDJRY-UHFFFAOYSA-N
Synonym 3-bromo-2-chloro-6-fluorotoluene, 2-chloro-3-bromo-6-fluorotoluene, 3-bromo-2-chlor-6-fluorotoluene, benzene, 1-bromo-2-chloro-4-fluoro-3-methyl, castest, pubchem4514, 1-bromo-2-chloro-4-fluoro-3-methyl-benzene, acmc-1ce0p, 3-bromo-2-chloro-6-fiuorotoluene, 3-bromo-2-chloro-6-fluoro-toluene
PubChem CID 2736227
IUPAC Name 1-bromo-2-chloro-4-fluoro-3-methylbenzene
SMILES CC1=C(C=CC(=C1Cl)Br)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15419898 SDS View Documents Alfa Aesar™
H63276.03
1g 36.50 €
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 21-08-2019
Please to purchase this item. Need a web account? Register with us today!
15429898 SDS View Documents Alfa Aesar™
H63276.06
5g 136.00 €
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 21-08-2019
Please to purchase this item. Need a web account? Register with us today!
15439898 SDS View Documents Alfa Aesar™
H63276.14
25g 550.00 €
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 21-08-2019
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

203302-92-5
223.469
VHQALVHKNXDJRY-UHFFFAOYSA-N
2736227
CC1=C(C=CC(=C1Cl)Br)F
C7H5BrClF
MFCD00236591
3-bromo-2-chloro-6-fluorotoluene, 2-chloro-3-bromo-6-fluorotoluene, 3-bromo-2-chlor-6-fluorotoluene, benzene, 1-bromo-2-chloro-4-fluoro-3-methyl, castest, pubchem4514, 1-bromo-2-chloro-4-fluoro-3-methyl-benzene, acmc-1ce0p, 3-bromo-2-chloro-6-fiuorotoluene, 3-bromo-2-chloro-6-fluoro-toluene
1-bromo-2-chloro-4-fluoro-3-methylbenzene

Specifications

3-Bromo-2-chloro-6-fluorotoluene
203302-92-5
MFCD00236591
VHQALVHKNXDJRY-UHFFFAOYSA-N
1-bromo-2-chloro-4-fluoro-3-methylbenzene
2736227
Solid
1g
C7H5BrClF
3-bromo-2-chloro-6-fluorotoluene, 2-chloro-3-bromo-6-fluorotoluene, 3-bromo-2-chlor-6-fluorotoluene, benzene, 1-bromo-2-chloro-4-fluoro-3-methyl, castest, pubchem4514, 1-bromo-2-chloro-4-fluoro-3-methyl-benzene, acmc-1ce0p, 3-bromo-2-chloro-6-fiuorotoluene, 3-bromo-2-chloro-6-fluoro-toluene
CC1=C(C=CC(=C1Cl)Br)F
223.469
223.47
97%