missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2,6-dimethylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60551.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475836

  • 646.00 € / 1g

Estimated Shipment date
from Supplier 17-09-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

941123-32-6
340.24
HJILQMNWGAAWMN-UHFFFAOYSA-N
8504710
C14H14BrNO2S
MFCD07957640
3-bromo-n-2,6-dimethylphenyl benzenesulfonamide, 3-bromo-n-2,6-dimethyl-phenyl-benzenesulfonamide, 3-bromo-n-2,6-dimethylphenyl benzene-1-sulfonamide

Specifications

3-Bromo-N-(2,6-dimethylphenyl)benzenesulfonamide
941123-32-6
MFCD07957640
HJILQMNWGAAWMN-UHFFFAOYSA-N
8504710
97%
1g
C14H14BrNO2S
3-bromo-n-2,6-dimethylphenyl benzenesulfonamide, 3-bromo-n-2,6-dimethyl-phenyl-benzenesulfonamide, 3-bromo-n-2,6-dimethylphenyl benzene-1-sulfonamide
340.24
340.23
Videos
SDS Documents