missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(3-chloro-4-methoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64323.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475675

  • 139.00 € / 1g


Estimated Shipment date
from Supplier 23-05-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665271

Specifications

3-Bromo-N-(3-chloro-4-methoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665271
Solid
1g
444.65
97%
Videos
SDS