missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide, 97%, Alfa Aesar™

Brand:  Alfa Aesar H64720.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15475705

  • 578.00 € / 5g


Estimated Shipment date
from Supplier 22-05-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665308

Specifications

3-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
MFCD27665308
Solid
5g
466.17
97%
Videos
SDS