missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-1-ol, 97%, Thermo Scientific Chemicals

Catalog Number p-80815
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
927-74-2
C4H6O
70.09
MFCD00002955
OTJZCIYGRUNXTP-UHFFFAOYSA-N
3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
13566
CHEBI:27444
but-3-yn-1-ol
C#CCCO
This item is not returnable. View return policy
927-74-2
C4H6O
70.09
MFCD00002955
OTJZCIYGRUNXTP-UHFFFAOYSA-N
3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
13566
CHEBI:27444
but-3-yn-1-ol
C#CCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -63.6°C
Color Colorless to Yellow
Density 0.9260g/mL
Boiling Point 128.8°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2CH2OH
Refractive Index 1.4397 to 1.4417
Quantity 5 mL
Beilstein 01, IV, 2219
Specific Gravity 0.926
Solubility Information Solubility in water: miscible
Formula Weight 70.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Butyn-1-ol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Butyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'