missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-5-nitrophenylboronic acid, 97%, Thermo Scientific™

Catalog Number 15264754
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
101084-81-5
C7H6BNO6
210.94
WNIFCLWDGNHGMX-UHFFFAOYSA-N
3-carboxy-5-nitrophenylboronic acid, 3-nitro-5-carboxyphenylboronic acid, 5-nitro-3-carboxyphenylboronic acid, 3-carboxy-5-nitrophenyl boronic acid, 3-carboxy-5-nitrobenzeneboronic acid, benzoic acid, 3-borono-5-nitro, 3-dihydroxyboryl-5-nitrobenzoic acid, 3-dihydroxyboranyl-5-nitrobenzoic acid, 3-borono-5-nitro-benzoic acid, pubchem5139
2773315
3-borono-5-nitrobenzoic acid
B(C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O)(O)O
This item is not returnable. View return policy
101084-81-5
C7H6BNO6
210.94
WNIFCLWDGNHGMX-UHFFFAOYSA-N
3-carboxy-5-nitrophenylboronic acid, 3-nitro-5-carboxyphenylboronic acid, 5-nitro-3-carboxyphenylboronic acid, 3-carboxy-5-nitrophenyl boronic acid, 3-carboxy-5-nitrobenzeneboronic acid, benzoic acid, 3-borono-5-nitro, 3-dihydroxyboryl-5-nitrobenzoic acid, 3-dihydroxyboranyl-5-nitrobenzoic acid, 3-borono-5-nitro-benzoic acid, pubchem5139
2773315
3-borono-5-nitrobenzoic acid
B(C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O)(O)O
Melting Point 225°C to 227°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 210.94
Percent Purity 97%
Chemical Name or Material 3-Carboxy-5-nitrophenylboronic acid
compliance-icons
Product Identifier
  • 3-Carboxy-5-nitrophenylboronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'