Learn More
3-Chloro-2-methylpropene, 98%
Description
3-Chloro-2-methylpropene has been used as reactant in the synthesis of cyclobutanone. It is used as an intermediate in the production of plastics, pharmaceuticals and other organic chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Chloro-2-methylpropene has been used as reactant in the synthesis of cyclobutanone. It is used as an intermediate in the production of plastics, pharmaceuticals and other organic chemicals.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -80°C |
| Density | 0.917 |
| Boiling Point | 71°C to 72°C |
| Flash Point | −12°C (9°F) |
| Refractive Index | 1.4275 |
| Quantity | 1000 g |
| UN Number | UN2554 |
| Merck Index | 14,2148 |
| Solubility Information | Not miscible in water. |
| Formula Weight | 90.55 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.