missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-3-methyl-1-butyne, 96%, Thermo Scientific Chemicals

Catalog Number p-3211880
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1111-97-3
C5H7Cl
102.56
MFCD00190221
QSILYWCNPOLKPN-UHFFFAOYSA-N
3-chloro-3-methyl-1-butyne, 1-butyne, 3-chloro-3-methyl, 3-chloro-3-methylbutyne, 3-methyl-3-chlorobutyne, acmc-1bo5c, 2-chloro-2-methyl butyne, 3-chloro-3-methyl butyne, 3-chloro-3-methyl-butyne, 3-chloro-3-methyl-1-butyn, 2-chloro-2-methyl-3-butyne
70679
3-chloro-3-methylbut-1-yne
CC(C)(Cl)C#C
This item is not returnable. View return policy
1111-97-3
C5H7Cl
102.56
MFCD00190221
QSILYWCNPOLKPN-UHFFFAOYSA-N
3-chloro-3-methyl-1-butyne, 1-butyne, 3-chloro-3-methyl, 3-chloro-3-methylbutyne, 3-methyl-3-chlorobutyne, acmc-1bo5c, 2-chloro-2-methyl butyne, 3-chloro-3-methyl butyne, 3-chloro-3-methyl-butyne, 3-chloro-3-methyl-1-butyn, 2-chloro-2-methyl-3-butyne
70679
3-chloro-3-methylbut-1-yne
CC(C)(Cl)C#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Straw
Density 0.9100g/mL
Boiling Point 73°C to 75°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4170 to 1.419
Quantity 1 g
Specific Gravity 0.91
Formula Weight 102.56
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Chloro-3-methyl-1-butyne
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Chloro-3-methyl-1-butyne
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'