missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-methylbenzylamine, 97%, Thermo Scientific™

Catalog Number p-2700006
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
67952-93-6
C8H10ClN
155.625
MFCD00014816
MXIGALIASISPNU-UHFFFAOYSA-N
3-chloro-4-methylbenzylamine, 3-chloro-4-methylphenyl methanamine, 1-3-chloro-4-methylphenyl methanamine, benzenemethanamine, 3-chloro-4-methyl, 3-chloro-4-methyl benzylamine, 3-chloro-4-methylphenyl methylamine, 3-chloro-4-methyl-benzylamine, rarechem al bw 1348, 4-aminomethyl-2-chlorotoluene, 3-chloro-4-methylbenzenemethanamine
106225
(3-chloro-4-methylphenyl)methanamine
CC1=C(C=C(C=C1)CN)Cl
This item is not returnable. View return policy
67952-93-6
C8H10ClN
155.625
MFCD00014816
MXIGALIASISPNU-UHFFFAOYSA-N
3-chloro-4-methylbenzylamine, 3-chloro-4-methylphenyl methanamine, 1-3-chloro-4-methylphenyl methanamine, benzenemethanamine, 3-chloro-4-methyl, 3-chloro-4-methyl benzylamine, 3-chloro-4-methylphenyl methylamine, 3-chloro-4-methyl-benzylamine, rarechem al bw 1348, 4-aminomethyl-2-chlorotoluene, 3-chloro-4-methylbenzenemethanamine
106225
(3-chloro-4-methylphenyl)methanamine
CC1=C(C=C(C=C1)CN)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 155.63
Percent Purity 97%
Chemical Name or Material 3-chloro-4-methylbenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'