missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzophenone, 98+%, Thermo Scientific Chemicals

Catalog Number p-7026831
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1016-78-0
C13H9ClO
216.664
MFCD00009816
CPLWKNRPZVNELG-UHFFFAOYSA-N
3-chlorobenzophenone, 3-chlorophenyl phenyl methanone, benzophenone, 3-chloro, methanone, 3-chlorophenyl phenyl, m-chlorobenzophenone, 3-chlorophenyl-phenylmethanone, methanone, 3-chlorophenyl pheny, 3-chlorophenyl phenyl ketone, 5-chlorobenzophenone, pubchem24414
66098
(3-chlorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Cl
This item is not returnable. View return policy
1016-78-0
C13H9ClO
216.664
MFCD00009816
CPLWKNRPZVNELG-UHFFFAOYSA-N
3-chlorobenzophenone, 3-chlorophenyl phenyl methanone, benzophenone, 3-chloro, methanone, 3-chlorophenyl phenyl, m-chlorobenzophenone, 3-chlorophenyl-phenylmethanone, methanone, 3-chlorophenyl pheny, 3-chlorophenyl phenyl ketone, 5-chlorobenzophenone, pubchem24414
66098
(3-chlorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 81°C to 85°C
Quantity 5 g
Beilstein 1869822
Formula Weight 216.67
Percent Purity ≥98%
Chemical Name or Material 3-Chlorobenzophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'