missing translation for 'onlineSavingsMsg'
Learn More

3-chlorophenyl isocyanate, 98%, ACROS Organics™

Brand:  Acros Organics 404965000

Code : 29

Additional Details : CAS Number : 2909-38-8 Weight : 0.55000kg Transport : UN number : 3382 Chem class : 6.1 Pack group : I


Packaging Glass bottle
Quantity 500g

 View more versions of this product

Product Code. 10424125

  • 93.61 € / 500g


Estimated Shipment
16-10-2018
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

2909-38-8
153.565
HHIRBXHEYVDUAM-UHFFFAOYSA-N
C7H4ClNO
MFCD00002016
17968

Specifications

3-Chlorophenyl isocyanate
82°C
Colorless to Yellow
500g
2909-38-8
100.0
1.5576 to 1.5596
98%
C7H4ClNO
MFCD00002016
C1=CC(=CC(=C1)Cl)N=C=O
153.565
153.57
1.2700g/mL
201.0°C
-4.0°C
98%
97.5
Authentic
Glass bottle
98%
NCOC6H4Cl
HHIRBXHEYVDUAM-UHFFFAOYSA-N
1-chloro-3-isocyanatobenzene
17968
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Chlorophenyl isocyanate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS