missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothieno[2,3-b]thiophene-2-carboxylic acid, 97%, Maybridge™

Brand:  Maybridge MO08116DA

Code : 31

Additional Details : CAS Number : 39244-08-1 Weight : 0.05100kg

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10002404

  • 163.00 € / 1g

Estimated Shipment
01-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

39244-08-1
218.669
GCQFYYGUNYJMNY-UHFFFAOYSA-N
4962405
C1=CSC2=C1C(=C(S2)C(=O)O)Cl
C7H3ClO2S2
MFCD01927113
3-chlorothieno 2,3-b thiophene-2-carboxylic acid, 4-chlorothieno 2,3-b thiophene-5-carboxylic acid, 3-chlorothiopheno 2,3-b thiophene-2-carboxylic acid, thieno 2,3-b thiophene-2-carboxylicacid, 3-chloro
4-chlorothieno[2,3-b]thiophene-5-carboxylic acid

Specifications

3-Chlorothieno[2, 3-b]thiophene-2-carboxylic acid
1g
97.0
C7H3ClO2S2
3-chlorothieno 2,3-b thiophene-2-carboxylic acid, 4-chlorothieno 2,3-b thiophene-5-carboxylic acid, 3-chlorothiopheno 2,3-b thiophene-2-carboxylic acid, thieno 2,3-b thiophene-2-carboxylicacid, 3-chloro
C1=CSC2=C1C(=C(S2)C(=O)O)Cl
218.669
218.68
Amber Glass Bottle
39244-08-1
97%
MFCD01927113
GCQFYYGUNYJMNY-UHFFFAOYSA-N
4-chlorothieno[2,3-b]thiophene-5-carboxylic acid
4962405
97%
Videos
SDS Documents