Learn More
3-(Dimethylamino)phenylboronic acid pinacol ester, 97%, ACROS Organics™
Brand: Acros Organics 444950010
Code : 29
Additional Details : CAS Number : 325142-87-8 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
325142-87-8 | |
247.14 | |
n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-n,n-dimethylamino phenylboronic acid, pinacol ester, 3-n,n-dimethylamino phenylboronic acid pinacol ester, dimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, 3-dimethylamino phenylboronic acid pinacol ester, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline, n,n-dimethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine | |
N,N-dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
C14H22BNO2 | |
BIJRPONWUBCGES-UHFFFAOYSA-N | |
17998909 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N(C)C |
Specifications
3-(Dimethylamino)phenylboronic acid pinacol ester | |
53°C to 55°C | |
325142-87-8 | |
C14H22BNO2 | |
BIJRPONWUBCGES-UHFFFAOYSA-N | |
N,N-dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
17998909 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-n,n-dimethylamino phenylboronic acid, pinacol ester, 3-n,n-dimethylamino phenylboronic acid pinacol ester, dimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, 3-dimethylamino phenylboronic acid pinacol ester, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline, n,n-dimethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N(C)C | |
247.14 | |
247.14 |
Safety and Handling
- 3-(Dimethylamino)phenylboronic acid pinacol ester
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.