Learn More
3-Dimethylaminopropiophenone Hydrochloride 99%, ACROS Organics™
Brand: Acros Organics 115890050
Code : 29
Additional Details : CAS Number : 879-72-1 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
879-72-1 | |
213.705 | |
DKNDBIIKSJWQFL-UHFFFAOYSA-N | |
92844 | |
CN(C)CCC(=O)C1=CC=CC=C1.Cl |
C11H16ClNO | |
MFCD00012481 | |
3-dimethylaminopropiophenone hydrochloride, 3-dimethylamino-1-phenylpropan-1-one hydrochloride, 3-dimethylamino propiophenone hydrochloride, beta-dimethylaminopropiophenone hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride, nsc 89, dimethylaminoethylphenylketone hydrochloride, propiophenone, 3-dimethylamino-, hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride 1:1, wln: 1n1&2vr &gh | |
3-(dimethylamino)-1-phenylpropan-1-one;hydrochloride |
Specifications
3-Dimethylaminopropiophenone hydrochloride | |
152.0°C to 155.0°C | |
99% | |
98.5 | |
99% | |
C6H5COCH2CH2N(CH3)2·HCl | |
14, II, 38 | |
DKNDBIIKSJWQFL-UHFFFAOYSA-N | |
3-(dimethylamino)-1-phenylpropan-1-one;hydrochloride | |
92844 | |
99% |
Glass bottle | |
5g | |
879-72-1 | |
100.0 | |
C11H16ClNO | |
MFCD00012481 | |
3-dimethylaminopropiophenone hydrochloride, 3-dimethylamino-1-phenylpropan-1-one hydrochloride, 3-dimethylamino propiophenone hydrochloride, beta-dimethylaminopropiophenone hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride, nsc 89, dimethylaminoethylphenylketone hydrochloride, propiophenone, 3-dimethylamino-, hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride 1:1, wln: 1n1&2vr &gh | |
CN(C)CCC(=O)C1=CC=CC=C1.Cl | |
213.705 | |
213.71 |
Safety and Handling
- 3-Dimethylaminopropiophenone hydrochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /