missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-formylphenylboronic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 167.93 |
Percent Purity | 95% |
Chemical Name or Material | 3-Fluoro-4-formylphenylboronic acid |
Safety and Handling
Product Identifier
- 3-Fluoro-4-formylphenylboronic acid
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'