Learn More
3-Fluoro-4-formylphenylboronic acid, 95%, ACROS Organics™
Brand: Acros Organics 445600050
Code : 29
Additional Details : CAS Number : 248270-25-9 Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
248270-25-9 | |
167.93 | |
3-fluoro-4-formylbenzeneboronic acid, 3-fluoro-4-formylphenyl boronic acid, 3-fluoro-4-formylphenboronic acid, boronic acid, 3-fluoro-4-formylphenyl, pubchem1847, acmc-1cga0, ksc494k4h, 4-borono-3-fluorobenzaldehyde, 3-fluoro-4-formyl-phenyl boronic acid, 3-fluoranyl-4-methanoyl-phenyl boronic acid | |
(3-fluoro-4-formylphenyl)boronic acid |
C7H6BFO3 | |
NZNRMUVHUVCIBR-UHFFFAOYSA-N | |
2778654 | |
B(C1=CC(=C(C=C1)C=O)F)(O)O |
Specifications
3-Fluoro-4-formylphenylboronic acid | |
248270-25-9 | |
C7H6BFO3 | |
NZNRMUVHUVCIBR-UHFFFAOYSA-N | |
(3-fluoro-4-formylphenyl)boronic acid | |
2778654 | |
95% |
5g | |
94% min. (HPLC) | |
3-fluoro-4-formylbenzeneboronic acid, 3-fluoro-4-formylphenyl boronic acid, 3-fluoro-4-formylphenboronic acid, boronic acid, 3-fluoro-4-formylphenyl, pubchem1847, acmc-1cga0, ksc494k4h, 4-borono-3-fluorobenzaldehyde, 3-fluoro-4-formyl-phenyl boronic acid, 3-fluoranyl-4-methanoyl-phenyl boronic acid | |
B(C1=CC(=C(C=C1)C=O)F)(O)O | |
167.93 | |
167.93 |
Safety and Handling
- 3-Fluoro-4-formylphenylboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.